Assay Details
| Gene Symbol: atp6v1g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079372.1 | NP_001072840.1 | 2 - 3 | 327 | 88 | |
| GenBank mRNA | BC121968.1 | - | 2 - 3 | 327 | 88 | |
| BC135509.1 | - | 2 - 3 | 282 | 88 | ||
More Information
Set Membership: |
|

atp6v1g2
Genomic Map