Assay Details
| Gene Symbol: nsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012966091.1 | XP_012821545.1 | 4 - 5 | 690 | 81 | |
| GenBank mRNA | BC122904.1 | - | 3 - 4 | 383 | 81 | |
| BC167929.1 | - | 4 - 5 | 643 | 81 | ||
More Information
Set Membership: |
|

nsg1
Genomic Map