Assay Details
| Gene Symbol: elf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079386.1 | NP_001072854.1 | 9 - 10 | 1569 | 70 | |
| XM_012957328.2 | XP_012812782.1 | 9 - 10 | 1599 | 70 | ||
| XM_012957329.2 | XP_012812783.1 | 8 - 9 | 1451 | 70 | ||
| XM_012957330.2 | XP_012812784.1 | 8 - 9 | 1573 | 70 | ||
| XM_012957331.2 | XP_012812785.1 | 8 - 9 | 1581 | 70 | ||
| GenBank mRNA | BC122943.1 | - | 9 - 10 | 1569 | 70 | |
More Information
Set Membership: |
|

elf1
Genomic Map