Assay Details
| Gene Symbol: med29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079404.1 | NP_001072872.1 | 3 - 4 | 372 | 93 | |
| GenBank mRNA | BC124010.1 | - | 3 - 4 | 372 | 93 | |
| BC135891.1 | - | 3 - 4 | 358 | 93 | ||
More Information
Set Membership: |
|

med29
Genomic Map