Assay Details
Gene Symbol: zmynd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079410.1 | NP_001072878.1 | 3 - 4 | 309 | 49 | |
GenBank mRNA | BC124055.1 | - | 3 - 4 | 309 | 49 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
