Assay Details
| Gene Symbol: egfl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079441.1 | NP_001072909.1 | 3 - 4 | 401 | 50 | |
| XM_012968703.1 | XP_012824157.1 | 2 - 3 | 412 | 50 | ||
| XM_012968704.2 | XP_012824158.1 | 2 - 3 | 528 | 50 | ||
| XM_018089875.1 | XP_017945364.1 | 2 - 3 | 708 | 50 | ||
| GenBank mRNA | BC123101.1 | - | 3 - 4 | 401 | 50 | |
More Information
Set Membership: |
|

egfl7
Genomic Map