Assay Details
| Gene Symbol: mindy4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079472.1 | NP_001072940.1 | 10 - 11 | 1622 | 71 | |
| GenBank mRNA | BC121261.1 | - | 10 - 11 | 1622 | 71 | |
More Information
Set Membership: |
|

mindy4
Genomic Map