Assay Details
| Gene Symbol: mindy4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079472.1 | NP_001072940.1 | 3 - 4 | 531 | 88 | |
| GenBank mRNA | BC121261.1 | - | 3 - 4 | 531 | 88 | |
| CU025041.1 | - | 3 - 4 | 535 | 88 | ||
More Information
Set Membership: |
|

mindy4
Genomic Map