Assay Details
| Gene Symbol: eef1a1o | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203970.3 | NP_989301.1 | 4 - 5 | 902 | 67 | |
| GenBank mRNA | BC064177.1 | - | 4 - 5 | 849 | 67 | |
| BC157320.1 | - | 4 - 5 | 810 | 67 | ||
| BC158425.1 | - | 4 - 5 | 806 | 67 | ||
| CR761806.2 | - | 4 - 5 | 776 | 67 | ||
More Information
Set Membership: |
|

eef1a1o
Genomic Map