Assay Details
| Gene Symbol: hexd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015910.3 | NP_001015910.1 | 8 - 9 | 955 | 72 | |
| XM_012971734.2 | XP_012827188.1 | 11 - 12 | 1283 | 72 | ||
| XM_012971735.2 | XP_012827189.1 | 11 - 12 | 1278 | 72 | ||
| XM_018097731.1 | XP_017953220.1 | 9 - 10 | 992 | 72 | ||
| XM_018097732.1 | XP_017953221.1 | 11 - 12 | 1222 | 72 | ||
| XM_018097733.1 | XP_017953222.1 | 10 - 11 | 1157 | 72 | ||
| GenBank mRNA | BC118693.1 | - | 8 - 9 | 955 | 72 | |
| CR926427.2 | - | 11 - 12 | 1227 | 72 | ||
More Information
Set Membership: |
|

hexd
Genomic Map