Assay Details
| Gene Symbol: hexd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015910.3 | NP_001015910.1 | 9 - 10 | 1041 | 77 | |
| XM_012971734.2 | XP_012827188.1 | 12 - 13 | 1369 | 77 | ||
| XM_012971735.2 | XP_012827189.1 | 12 - 13 | 1364 | 77 | ||
| XM_018097731.1 | XP_017953220.1 | 10 - 11 | 1078 | 77 | ||
| XM_018097732.1 | XP_017953221.1 | 12 - 13 | 1308 | 77 | ||
| XM_018097733.1 | XP_017953222.1 | 11 - 12 | 1243 | 77 | ||
| GenBank mRNA | BC118693.1 | - | 9 - 10 | 1041 | 77 | |
| CR926427.2 | - | 12 - 13 | 1313 | 77 | ||
More Information
Set Membership: |
|

hexd
Genomic Map