Assay Details
| Gene Symbol: capsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016524.3 | NP_001016524.1 | 2 - 3 | 305 | 95 | |
| XM_012962427.2 | XP_012817881.1 | 3 - 4 | 734 | 95 | ||
| XM_012962430.2 | XP_012817884.1 | 3 - 4 | 742 | 95 | ||
| GenBank mRNA | BC121608.1 | - | 2 - 3 | 305 | 95 | |
| CR926201.2 | - | 3 - 4 | 341 | 95 | ||
More Information
Set Membership: |
|

capsl
Genomic Map