Assay Details
| Gene Symbol: rps24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016625.3 | NP_001016625.1 | 2 - 3 | 91 | 75 | |
| XM_012966613.2 | XP_012822067.1 | 2 - 3 | 75 | |||
| XM_012966614.2 | XP_012822068.1 | 2 - 3 | 75 | |||
| GenBank mRNA | BC135566.1 | - | 2 - 3 | 91 | 75 | |
| CR760480.2 | - | 1 - 2 | 75 | |||
More Information
Set Membership: |
|

rps24
Genomic Map