Assay Details
| Gene Symbol: borcs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016829.3 | NP_001016829.1 | 4 - 5 | 295 | 93 | |
| GenBank mRNA | BC091604.1 | - | 4 - 5 | 295 | 93 | |
| CR855513.2 | - | 4 - 5 | 423 | 93 | ||
More Information
Set Membership: |
|

borcs7
Genomic Map