Assay Details
| Gene Symbol: plpp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016858.4 | NP_001016858.3 | 2 - 3 | 628 | 97 | |
| GenBank mRNA | BC135252.1 | - | 2 - 3 | 628 | 97 | |
More Information
Set Membership: |
|

plpp3
Genomic Map