Assay Details
| Gene Symbol: plpp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016858.4 | NP_001016858.3 | 5 - 6 | 1144 | 90 | |
| XM_012961143.2 | XP_012816597.1 | 5 - 6 | 1777 | 90 | ||
| GenBank mRNA | AL635807.2 | - | 2 - 3 | 538 | 90 | |
| BC135252.1 | - | 5 - 6 | 1144 | 90 | ||
More Information
Set Membership: |
|

plpp3
Genomic Map