Assay Details
| Gene Symbol: tpm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017087.3 | NP_001017087.1 | 3 - 4 | 698 | 75 | |
| XM_012963258.2 | XP_012818712.1 | 4 - 5 | 650 | 75 | ||
| XM_012963262.2 | XP_012818716.1 | 4 - 5 | 652 | 75 | ||
| XM_012963264.2 | XP_012818718.1 | 3 - 4 | 889 | 75 | ||
| XM_012963267.2 | XP_012818721.1 | 4 - 5 | 726 | 75 | ||
| XM_012963269.2 | XP_012818723.1 | 3 - 4 | 894 | 75 | ||
| GenBank mRNA | BC135407.1 | - | 3 - 4 | 698 | 75 | |
| CR761414.2 | - | 3 - 4 | 554 | 75 | ||
More Information
Set Membership: |
|

tpm4
Genomic Map