Assay Details
| Gene Symbol: pan3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017301.3 | NP_001017301.1 | 11 - 12 | 1773 | 58 | |
| XM_012957062.2 | XP_012812516.1 | 11 - 12 | 2107 | 58 | ||
| XM_018091283.1 | XP_017946772.1 | 11 - 12 | 1378 | 58 | ||
| GenBank mRNA | BC128994.1 | - | 11 - 12 | 1773 | 58 | |
| CR855590.2 | - | 11 - 12 | 1711 | 58 | ||
More Information
Set Membership: |
|

pan3
Genomic Map