Assay Details
| Gene Symbol: nrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032306.2 | NP_001027477.2 | 2 - 3 | 546 | 76 | |
| XM_012953266.2 | XP_012808720.1 | 4 - 5 | 700 | 76 | ||
| XM_012953267.2 | XP_012808721.1 | 4 - 5 | 670 | 76 | ||
| XM_012953268.1 | XP_012808722.1 | 3 - 4 | 599 | 76 | ||
| GenBank mRNA | BC135756.1 | - | 2 - 3 | 546 | 76 | |
| DQ018733.1 | - | 1 - 2 | 430 | 76 | ||
More Information
Set Membership: |
|

nrl
Genomic Map