Assay Details
| Gene Symbol: tmem222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078800.2 | NP_001072268.2 | 4 - 5 | 340 | 78 | |
| GenBank mRNA | BC121266.1 | - | 4 - 5 | 340 | 78 | |
| CU075502.1 | - | 4 - 5 | 329 | 78 | ||
More Information
Set Membership: |
|

tmem222
Genomic Map