Assay Details
| Gene Symbol: pcdh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079015.1 | NP_001072483.1 | 3 - 4 | 3169 | 82 | |
| XM_012965336.2 | XP_012820790.1 | 3 - 4 | 3202 | 82 | ||
| GenBank mRNA | BC121910.1 | - | 3 - 4 | 3169 | 82 | |
More Information
Set Membership: |
|

pcdh10
Genomic Map