Assay Details
| Gene Symbol: kcnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079016.1 | NP_001072484.1 | 4 - 5 | 1052 | 63 | |
| XM_012965355.2 | XP_012820809.1 | 4 - 5 | 1040 | 63 | ||
| XM_012965371.2 | XP_012820825.1 | 4 - 5 | 1040 | 63 | ||
| GenBank mRNA | BC121911.1 | - | 4 - 5 | 1052 | 63 | |
More Information
Set Membership: |
|

kcnn1
Genomic Map