Assay Details
| Gene Symbol: scamp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079024.1 | NP_001072492.1 | 4 - 5 | 674 | 90 | |
| XM_018092169.1 | XP_017947658.1 | 5 - 6 | 584 | 90 | ||
| GenBank mRNA | BC121923.1 | - | 4 - 5 | 674 | 90 | |
More Information
Set Membership: |
|

scamp5
Genomic Map