Assay Details
| Gene Symbol: rbm46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079033.1 | NP_001072501.1 | 2 - 3 | 243 | 84 | |
| XM_012965392.2 | XP_012820846.1 | 2 - 3 | 290 | 84 | ||
| GenBank mRNA | BC121934.1 | - | 2 - 3 | 243 | 84 | |
| Gene Symbol: LOC100498620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018093696.1 | XP_017949185.1 | 1 - 2 | 214 | 84 | |
More Information
Set Membership: |
|

rbm46
Genomic Map