Assay Details
| Gene Symbol: gpr83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079037.1 | NP_001072505.1 | 3 - 4 | 915 | 92 | |
| GenBank mRNA | BC121939.1 | - | 3 - 4 | 915 | 92 | |
More Information
Set Membership: |
|

gpr83
Genomic Map