Assay Details
| Gene Symbol: fbxo31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079057.1 | NP_001072525.1 | 8 - 9 | 1100 | 62 | |
| XM_012961337.2 | XP_012816791.1 | 7 - 8 | 1329 | 62 | ||
| XM_012961338.2 | XP_012816792.1 | 8 - 9 | 868 | 62 | ||
| XM_018093094.1 | XP_017948583.1 | 7 - 8 | 781 | 62 | ||
| GenBank mRNA | BC121976.1 | - | 8 - 9 | 1100 | 62 | |
More Information
Set Membership: |
|

fbxo31
Genomic Map