Assay Details
| Gene Symbol: chst8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079061.1 | NP_001072529.1 | 3 - 4 | 742 | 81 | |
| XM_018093095.1 | XP_017948584.1 | 4 - 5 | 676 | 81 | ||
| GenBank mRNA | BC121984.1 | - | 3 - 4 | 742 | 81 | |
More Information
Set Membership: |
|

chst8
Genomic Map