Assay Details
| Gene Symbol: katnbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079080.1 | NP_001072548.1 | 9 - 10 | 961 | 71 | |
| XM_012968623.2 | XP_012824077.1 | 8 - 9 | 983 | 71 | ||
| XM_018096320.1 | XP_017951809.1 | 7 - 8 | 932 | 71 | ||
| GenBank mRNA | BC122056.1 | - | 9 - 10 | 961 | 71 | |
| CT030061.2 | - | 9 - 10 | 959 | 71 | ||
More Information
Set Membership: |
|

katnbl1
Genomic Map