Assay Details
| Gene Symbol: trim63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079089.1 | NP_001072557.1 | 3 - 4 | 540 | 71 | |
| XM_012957279.2 | XP_012812733.1 | 3 - 4 | 647 | 71 | ||
| GenBank mRNA | BC122073.1 | - | 3 - 4 | 540 | 71 | |
More Information
Set Membership: |
|

trim63
Genomic Map