Assay Details
| Gene Symbol: adat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079094.1 | NP_001072562.1 | 2 - 3 | 150 | 66 | |
| GenBank mRNA | BC122084.1 | - | 2 - 3 | 150 | 66 | |
More Information
Set Membership: |
|

adat2
Genomic Map