Assay Details
| Gene Symbol: gnaz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079121.1 | NP_001072589.1 | 1 - 2 | 1335 | 82 | |
| XM_012965554.2 | XP_012821008.1 | 2 - 3 | 1685 | 82 | ||
| GenBank mRNA | BC122955.1 | - | 1 - 2 | 1335 | 82 | |
More Information
Set Membership: |
|

gnaz
Genomic Map