Assay Details
| Gene Symbol: car1_predicted | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079317.1 | NP_001072785.1 | 3 - 4 | 355 | 98 | |
| XM_012964940.2 | XP_012820394.1 | 3 - 4 | 415 | 98 | ||
| XM_012964941.2 | XP_012820395.1 | 3 - 4 | 490 | 98 | ||
| XM_012964943.2 | XP_012820397.1 | 3 - 4 | 827 | 98 | ||
| XM_012964944.2 | XP_012820398.1 | 3 - 4 | 409 | 98 | ||
| XM_018094705.1 | XP_017950194.1 | 3 - 4 | 1374 | 98 | ||
| GenBank mRNA | BC118911.1 | - | 3 - 4 | 355 | 98 | |
More Information
Set Membership: |
|

car1_predicted
Genomic Map