Assay Details
| Gene Symbol: ube2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079423.1 | NP_001072891.1 | 4 - 5 | 314 | 98 | |
| GenBank mRNA | BC125770.1 | - | 4 - 5 | 314 | 98 | |
More Information
Set Membership: |
|

ube2d1
Genomic Map