Assay Details
| Gene Symbol: sgcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079440.1 | NP_001072908.1 | 6 - 7 | 671 | 66 | |
| XM_012957334.2 | XP_012812788.1 | 6 - 7 | 755 | 66 | ||
| XM_018091369.1 | XP_017946858.1 | 6 - 7 | 752 | 66 | ||
| GenBank mRNA | BC122992.1 | - | 6 - 7 | 671 | 66 | |
| BC160536.1 | - | 6 - 7 | 659 | 66 | ||
More Information
Set Membership: |
|

sgcg
Genomic Map