Assay Details
| Gene Symbol: klhl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079442.1 | NP_001072910.1 | 5 - 6 | 1274 | 61 | |
| XM_012966300.2 | XP_012821754.1 | 5 - 6 | 1321 | 61 | ||
| XM_012966303.2 | XP_012821757.1 | 5 - 6 | 1306 | 61 | ||
| XM_012966306.2 | XP_012821760.1 | 5 - 6 | 1261 | 61 | ||
| XM_012966310.2 | XP_012821764.1 | 5 - 6 | 1272 | 61 | ||
| XM_012966315.2 | XP_012821769.1 | 4 - 5 | 967 | 61 | ||
| GenBank mRNA | BC124036.1 | - | 5 - 6 | 1274 | 61 | |
More Information
Set Membership: |
|

klhl22
Genomic Map