Assay Details
| Gene Symbol: tox4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097155.1 | NP_001090624.1 | 7 - 8 | 1607 | 73 | |
| XM_012958612.2 | XP_012814066.1 | 8 - 9 | 1947 | 73 | ||
| XM_012958617.2 | XP_012814071.1 | 8 - 9 | 1819 | 73 | ||
| XM_012958625.2 | XP_012814079.1 | 8 - 9 | 1752 | 73 | ||
| GenBank mRNA | BC064865.1 | - | 7 - 8 | 1598 | 73 | |
| BC135950.1 | - | 7 - 8 | 1607 | 73 | ||
More Information
Set Membership: |
|

tox4
Genomic Map