Assay Details
| Gene Symbol: cyp19a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097161.1 | NP_001090630.1 | 9 - 10 | 1523 | 76 | |
| GenBank mRNA | AB244215.1 | - | 8 - 9 | 1366 | 76 | |
| BC118823.1 | - | 9 - 10 | 1523 | 76 | ||
More Information
Set Membership: |
|

cyp19a1
Genomic Map