Assay Details
| Gene Symbol: sall1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097177.1 | NP_001090646.1 | 3 - 4 | 3667 | 91 | |
| XM_012960358.2 | XP_012815812.1 | 4 - 5 | 4650 | 91 | ||
| XM_012960360.2 | XP_012815814.1 | 3 - 4 | 4560 | 91 | ||
| XM_018092857.1 | XP_017948346.1 | 2 - 3 | 3675 | 91 | ||
| GenBank mRNA | BC127274.1 | - | 3 - 4 | 3667 | 91 | |
| CU024990.1 | - | 5 - 6 | 1094 | 91 | ||
More Information
Set Membership: |
|

sall1
Genomic Map