Assay Details
| Gene Symbol: cnnm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097203.1 | NP_001090672.1 | 1 - 2 | 1060 | 68 | |
| XM_018091894.1 | XP_017947383.1 | 1 - 2 | 1553 | 68 | ||
| XM_018091895.1 | XP_017947384.1 | 1 - 2 | 1553 | 68 | ||
| GenBank mRNA | BC127319.1 | - | 1 - 2 | 1060 | 68 | |
More Information
Set Membership: |
|

cnnm2
Genomic Map