Assay Details
| Gene Symbol: tmeff2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097223.2 | NP_001090692.1 | 2 - 3 | 349 | 70 | |
| XM_012969926.2 | XP_012825380.1 | 2 - 3 | 569 | 70 | ||
| GenBank mRNA | BC127375.1 | - | 2 - 3 | 349 | 70 | |
More Information
Set Membership: |
|

tmeff2
Genomic Map