Assay Details
| Gene Symbol: etv3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097229.1 | NP_001090698.1 | 4 - 5 | 449 | 59 | |
| XM_012967807.2 | XP_012823261.1 | 4 - 5 | 587 | 59 | ||
| GenBank mRNA | BC128620.1 | - | 4 - 5 | 449 | 59 | |
| CT030495.1 | - | 3 - 4 | 427 | 59 | ||
More Information
Set Membership: |
|

etv3
Genomic Map