Assay Details
| Gene Symbol: zswim5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097236.1 | NP_001090705.1 | 1 - 2 | 580 | 57 | |
| GenBank mRNA | BC128630.1 | - | 1 - 2 | 580 | 57 | |
More Information
Set Membership: |
|

zswim5
Genomic Map