Assay Details
| Gene Symbol: arhgap21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097292.1 | NP_001090761.1 | 11 - 12 | 3051 | 56 | |
| XM_018094500.1 | XP_017949989.1 | 12 - 13 | 3173 | 56 | ||
| GenBank mRNA | BC133058.1 | - | 11 - 12 | 3051 | 56 | |
More Information
Set Membership: |
|

arhgap21
Genomic Map