Assay Details
| Gene Symbol: cyp3a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097314.1 | NP_001090783.1 | 10 - 11 | 1007 | 81 | |
| XM_012964140.2 | XP_012819594.1 | 9 - 10 | 1010 | 81 | ||
| XM_012964141.2 | XP_012819595.1 | 10 - 11 | 1067 | 81 | ||
| GenBank mRNA | BC135164.1 | - | 10 - 11 | 1007 | 81 | |
More Information
Set Membership: |
|

cyp3a5
Genomic Map