Assay Details
| Gene Symbol: clvs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097346.1 | NP_001090815.1 | 3 - 4 | 862 | 65 | |
| XM_012964142.2 | XP_012819596.1 | 3 - 4 | 877 | 65 | ||
| GenBank mRNA | BC135266.1 | - | 3 - 4 | 862 | 65 | |
More Information
Set Membership: |
|

clvs1
Genomic Map