Assay Details
| Gene Symbol: gem | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097353.1 | NP_001090822.1 | 2 - 3 | 410 | 71 | |
| GenBank mRNA | BC121659.1 | - | 2 - 3 | 410 | 71 | |
More Information
Set Membership: |
|

gem
Genomic Map