Assay Details
| Gene Symbol: h2afy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097360.1 | NP_001090829.1 | 8 - 9 | 1008 | 92 | |
| XM_012958366.2 | XP_012813820.1 | 8 - 9 | 1174 | 92 | ||
| GenBank mRNA | BC089240.1 | - | 8 - 9 | 1008 | 92 | |
More Information
Set Membership: |
|

h2afy
Genomic Map