Assay Details
| Gene Symbol: fus | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097367.1 | NP_001090836.1 | 8 - 9 | 912 | 88 | |
| XM_012953742.2 | XP_012809196.1 | 8 - 9 | 1012 | 88 | ||
| XM_012953743.2 | XP_012809197.1 | 8 - 9 | 1011 | 88 | ||
| GenBank mRNA | BC168544.1 | - | 8 - 9 | 913 | 88 | |
| CR848185.2 | - | 8 - 9 | 912 | 88 | ||
More Information
Set Membership: |
|

fus
Genomic Map