Assay Details
| Gene Symbol: paip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097384.1 | NP_001090853.1 | 1 - 2 | 309 | 60 | |
| GenBank mRNA | BC135141.1 | - | 1 - 2 | 309 | 60 | |
More Information
Set Membership: |
|

paip1
Genomic Map