Assay Details
| Gene Symbol: angptl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097403.1 | NP_001090872.1 | 4 - 5 | 1650 | 56 | |
| XM_012969751.2 | XP_012825205.1 | 4 - 5 | 1440 | 56 | ||
| GenBank mRNA | BC135324.1 | - | 4 - 5 | 1650 | 56 | |
More Information
Set Membership: |
|

angptl2
Genomic Map